Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009274.1 | NP_001009274.1 | 1 - 2 | 149 | 120 | |
GenBank mRNA | AY205232.1 | - | 1 - 2 | 149 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |