Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166204.1 | NP_001159676.1 | 1 - 2 | 145 | 91 | |
XM_012106119.3 | XP_011961509.1 | 2 - 3 | 272 | 91 | ||
XM_012106120.3 | XP_011961510.1 | 2 - 3 | 288 | 91 | ||
XM_012106121.3 | XP_011961511.1 | 2 - 3 | 272 | 91 | ||
GenBank mRNA | GQ221059.1 | - | 1 - 2 | 145 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |