Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DQA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308597.1 | NP_001295526.1 | 4 - 5 | 792 | 123 | |
GenBank mRNA | HG798791.1 | - | 4 - 5 | 792 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |