Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NPAS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287463.1 | NP_001274392.1 | 6 - 7 | 954 | 85 | |
GenBank mRNA | HF912412.1 | - | 6 - 7 | 954 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |