Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: GATD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162560.1 | NP_001156032.1 | 4 - 5 | 453 | 106 | |
GenBank mRNA | FJ969404.1 | - | 4 - 5 | 453 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |