Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134305.1 | NP_001127777.1 | 3 - 4 | 330 | 109 | |
XM_027965719.1 | XP_027821520.1 | 5 - 6 | 492 | 109 | ||
XM_027965720.1 | XP_027821521.1 | 4 - 5 | 516 | 109 | ||
GenBank mRNA | EU797610.1 | - | 3 - 4 | 330 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |