Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CYP2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001245972.1 | NP_001232901.1 | 3 - 4 | 506 | 80 | |
GenBank mRNA | HQ263378.1 | - | 3 - 4 | 506 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |