Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015096190.2 | XP_014951676.2 | 5 - 6 | 2957 | 72 | |
XM_027970507.1 | XP_027826308.1 | 5 - 6 | 515 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |