Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002387.4 | XP_004002436.2 | 6 - 7 | 1236 | 96 | |
XM_012179575.3 | XP_012034965.2 | 6 - 7 | 1213 | 96 | ||
XM_012179579.3 | XP_012034969.2 | 7 - 8 | 1319 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |