Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZAR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973019.1 | XP_027828820.1 | 2 - 3 | 1206 | 104 | |
XM_027973020.1 | XP_027828821.1 | 2 - 3 | 1206 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |