Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002768.3 | XP_004002817.2 | 2 - 3 | 1042 | 116 | |
XM_012186326.2 | XP_012041716.2 | 2 - 3 | 1054 | 116 | ||
XM_027978278.1 | XP_027834079.1 | 2 - 3 | 233 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |