Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101112936 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002644.4 | XP_004002693.2 | 2 - 3 | 464 | 130 | |
XM_027976821.1 | XP_027832622.1 | 2 - 3 | 466 | 130 | ||
GenBank mRNA | KX931094.1 | - | 2 - 3 | 466 | 130 | |
KX931095.1 | - | 2 - 3 | 466 | 130 | ||
KX931096.1 | - | 2 - 3 | 466 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |