Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094393.2 | XP_014949879.2 | 1 - 2 | 984 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |