Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGPEP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015101866.2 | XP_014957352.2 | 2 - 3 | 375 | 62 | |
XM_027957022.1 | XP_027812823.1 | 2 - 3 | 396 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |