Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNRD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012102131.2 | XP_011957521.1 | 2 - 3 | 402 | 78 | |
XM_012102132.2 | XP_011957522.1 | 3 - 4 | 318 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |