Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012185529.2 | XP_012040919.2 | 2 - 3 | 707 | 94 | |
XM_027975108.1 | XP_027830909.1 | 3 - 4 | 1234 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |