Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LRRC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027968779.1 | XP_027824580.1 | 2 | 2471 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |