Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: SLC36A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082183.1 | NP_001075652.1 | 9 - 10 | 1100 | 66 | |
XM_008254838.2 | XP_008253060.1 | 9 - 10 | 1179 | 66 | ||
XM_008254845.2 | XP_008253067.1 | 9 - 10 | 1294 | 66 | ||
XM_008254846.2 | XP_008253068.1 | 9 - 10 | 1213 | 66 | ||
XM_017340981.1 | XP_017196470.1 | 9 - 10 | 1190 | 66 | ||
GenBank mRNA | AY989816.1 | - | 9 - 10 | 1100 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |