Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082697.1 | NP_001076166.1 | 1361 | 72 | ||
GenBank mRNA | M87007.1 | - | 1361 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus |