Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081996.1 | NP_001075465.1 | 4 | 1551 | 77 | |
XM_008255637.2 | XP_008253859.1 | 4 | 1681 | 77 | ||
XM_008255638.2 | XP_008253860.1 | 4 | 1681 | 77 | ||
XM_008255639.2 | XP_008253861.1 | 4 | 1681 | 77 | ||
GenBank mRNA | AB023447.1 | - | 4 | 1551 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |