Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171288.1 | NP_001164759.1 | 3 - 4 | 421 | 86 | |
XM_008261390.2 | XP_008259612.1 | 5 - 6 | 651 | 86 | ||
XM_008261391.2 | XP_008259613.1 | 5 - 6 | 626 | 86 | ||
XM_008261392.2 | XP_008259614.1 | 5 - 6 | 622 | 86 | ||
XM_008261393.2 | XP_008259615.1 | 5 - 6 | 598 | 86 | ||
XM_008261394.2 | XP_008259616.1 | 4 - 5 | 1106 | 86 | ||
XM_017343656.1 | XP_017199145.1 | 5 - 6 | 1094 | 86 | ||
XM_017343657.1 | XP_017199146.1 | 5 - 6 | 1069 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |