Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BNIPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028829186.1 | XP_028685019.1 | 1030 | 128 | ||
GenBank mRNA | JU478220.1 | - | 806 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |