Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193479.2 | NP_001180408.1 | 509 | 77 | ||
GenBank mRNA | JU474687.1 | - | 487 | 77 | ||
JV054772.1 | - | 509 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |