Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266926.1 | NP_001253855.1 | 509 | 80 | ||
XM_015126952.1 | XP_014982438.1 | 410 | 80 | |||
XM_015126953.2 | XP_014982439.1 | 612 | 80 | |||
GenBank mRNA | JU320629.1 | - | 509 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |