Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266926.1 | NP_001253855.1 | 708 | 130 | ||
XM_015126952.1 | XP_014982438.1 | 609 | 130 | |||
GenBank mRNA | JU320629.1 | - | 708 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |