Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266745.1 | NP_001253674.1 | 483 | 116 | ||
XM_015126509.2 | XP_014981995.1 | 812 | 116 | |||
XM_015126513.2 | XP_014981999.1 | 549 | 116 | |||
XM_028844038.1 | XP_028699871.1 | 501 | 116 | |||
XM_028844041.1 | XP_028699874.1 | 1018 | 116 | |||
XM_028844042.1 | XP_028699875.1 | 1018 | 116 | |||
GenBank mRNA | JV045342.1 | - | 116 | |||
JV616465.1 | - | 542 | 116 | |||
JV667262.1 | - | 483 | 116 |
Gene Symbol: HEATR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU335232.1 | - | 9916 | 116 | ||
JU335233.1 | - | 9916 | 116 | |||
JU476199.1 | - | 9918 | 116 | |||
JV717936.1 | - | 9906 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |