Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028851802.1 | XP_028707635.1 | 1237 | 66 | ||
XM_028851803.1 | XP_028707636.1 | 1234 | 66 | |||
GenBank mRNA | JU470493.1 | - | 542 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |