Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198663.2 | NP_001185592.1 | 5138 | 153 | ||
GenBank mRNA | AY680468.1 | - | 460 | 153 | ||
JU322025.1 | - | 5179 | 153 | |||
JU475564.1 | - | 5100 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |