Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172082.1 | NP_001165553.1 | 229 | 121 | ||
GenBank mRNA | AF334809.1 | - | 135 | 121 | ||
CD766724.1 | - | 233 | 121 | |||
JU323048.1 | - | 248 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |