Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOBEC3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198693.2 | NP_001185622.1 | 1016 | 72 | ||
GenBank mRNA | AY331716.1 | - | 715 | 72 | ||
DQ913647.1 | - | 708 | 72 | |||
DQ913648.1 | - | 702 | 72 | |||
DQ913649.1 | - | 631 | 72 | |||
DQ913650.1 | - | 706 | 72 | |||
DQ913651.1 | - | 561 | 72 | |||
DQ913652.1 | - | 682 | 72 | |||
DQ913653.1 | - | 702 | 72 | |||
DQ913654.1 | - | 561 | 72 | |||
DQ913655.1 | - | 643 | 72 | |||
DQ913656.1 | - | 702 | 72 | |||
DQ913657.1 | - | 702 | 72 | |||
DQ913658.1 | - | 702 | 72 | |||
DQ913659.1 | - | 702 | 72 | |||
DQ913660.1 | - | 699 | 72 | |||
DQ913661.1 | - | 702 | 72 | |||
DQ913662.1 | - | 702 | 72 | |||
DQ913663.1 | - | 359 | 72 | |||
DQ913664.1 | - | 702 | 72 | |||
DQ913665.1 | - | 563 | 72 | |||
DQ913666.1 | - | 561 | 72 | |||
DQ913667.1 | - | 681 | 72 | |||
DQ913668.1 | - | 707 | 72 | |||
DQ913669.1 | - | 708 | 72 | |||
DQ913670.1 | - | 561 | 72 | |||
DQ913671.1 | - | 702 | 72 | |||
DQ913672.1 | - | 681 | 72 | |||
DQ913673.1 | - | 561 | 72 | |||
DQ913674.1 | - | 565 | 72 | |||
DQ913675.1 | - | 561 | 72 | |||
DQ913676.1 | - | 702 | 72 | |||
DQ913677.1 | - | 678 | 72 | |||
DQ913678.1 | - | 702 | 72 | |||
DQ913679.1 | - | 699 | 72 | |||
DQ913680.1 | - | 679 | 72 | |||
DQ913681.1 | - | 563 | 72 | |||
DQ913682.1 | - | 560 | 72 | |||
DQ913683.1 | - | 707 | 72 | |||
DQ913684.1 | - | 702 | 72 | |||
DQ913685.1 | - | 565 | 72 | |||
DQ913686.1 | - | 561 | 72 | |||
DQ913687.1 | - | 679 | 72 | |||
DQ913688.1 | - | 563 | 72 | |||
DQ913689.1 | - | 561 | 72 | |||
DQ913690.1 | - | 681 | 72 | |||
JU491021.1 | - | 726 | 72 | |||
JX915798.1 | - | 739 | 72 | |||
KC176183.1 | - | 736 | 72 | |||
KC176184.1 | - | 736 | 72 | |||
KF020481.1 | - | 739 | 72 | |||
KF020482.1 | - | 739 | 72 | |||
KF020483.1 | - | 739 | 72 | |||
KF020484.1 | - | 739 | 72 | |||
KF020485.1 | - | 739 | 72 | |||
KF020486.1 | - | 736 | 72 | |||
KU058146.1 | - | 943 | 72 | |||
KU058147.2 | - | 772 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta Inventoried |