Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266637.1 | NP_001253566.1 | 420 | 121 | ||
XM_015136475.2 | XP_014991961.2 | 512 | 121 | |||
GenBank mRNA | JU334141.1 | - | 394 | 121 | ||
JU473257.1 | - | 302 | 121 | |||
JV044100.1 | - | 420 | 121 | |||
JV639287.1 | - | 434 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |