Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194864.2 | NP_001181793.1 | 424 | 71 | ||
NM_001376986.1 | NP_001363915.1 | 462 | 71 | |||
XM_015136060.2 | XP_014991546.1 | 857 | 71 | |||
GenBank mRNA | JU322675.1 | - | 462 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta Inventoried |