Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015128086.2 | XP_014983572.1 | 680 | 97 | ||
GenBank mRNA | JU321943.1 | - | 150 | 97 | ||
JV704174.1 | - | 153 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |