Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266368.1 | NP_001253297.1 | 273 | 101 | ||
XM_015147132.2 | XP_015002618.1 | 369 | 101 | |||
GenBank mRNA | JU336485.1 | - | 273 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |