Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLX1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194449.2 | NP_001181378.1 | 450 | 158 | ||
GenBank mRNA | JU470693.1 | - | 158 | |||
JV635534.1 | - | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |