Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGMAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257877.1 | NP_001244806.1 | 222 | 83 | ||
XM_028834256.1 | XP_028690089.1 | 1374 | 83 | |||
GenBank mRNA | JU320933.1 | - | 222 | 83 | ||
JU472376.1 | - | 162 | 83 | |||
JV044654.1 | - | 153 | 83 | |||
JV530655.1 | - | 227 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |