Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194897.3 | NP_001181826.2 | 338 | 168 | ||
XM_015143992.2 | XP_014999478.1 | 655 | 168 | |||
GenBank mRNA | JU322906.1 | - | 331 | 168 | ||
JU475861.1 | - | 322 | 168 | |||
JV686060.1 | - | 338 | 168 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |