Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107477.4 | XP_001107477.2 | 711 | 136 | ||
XM_028826517.1 | XP_028682350.1 | 592 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |