Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144745.2 | XP_015000231.2 | 850 | 79 | ||
XM_015144746.2 | XP_015000232.2 | 984 | 79 | |||
XM_015144747.2 | XP_015000233.2 | 1103 | 79 | |||
XM_015144748.2 | XP_015000234.1 | 719 | 79 | |||
GenBank mRNA | JU333772.1 | - | 756 | 79 | ||
JU475016.1 | - | 759 | 79 | |||
JV046639.1 | - | 791 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |