Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110694.2 | XP_014966180.1 | 945 | 116 | ||
XM_015110695.2 | XP_014966181.1 | 1286 | 116 | |||
XM_028830963.1 | XP_028686796.1 | 1003 | 116 | |||
XM_028830964.1 | XP_028686797.1 | 1316 | 116 | |||
XM_028830965.1 | XP_028686798.1 | 698 | 116 | |||
XM_028830966.1 | XP_028686799.1 | 1285 | 116 | |||
XM_028830967.1 | XP_028686800.1 | 896 | 116 | |||
GenBank mRNA | JU332938.1 | - | 911 | 116 | ||
JU480359.1 | - | 618 | 116 | |||
JV046205.1 | - | 830 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |