Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RWDD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257574.1 | NP_001244503.1 | 222 | 96 | ||
XM_015139574.2 | XP_014995060.1 | 291 | 96 | |||
XM_015139575.2 | XP_014995061.1 | 285 | 96 | |||
GenBank mRNA | JU333319.1 | - | 222 | 96 | ||
JU474629.1 | - | 205 | 96 | |||
JV046409.1 | - | 216 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |