Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2AFX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194205.2 | NP_001181134.1 | 145 | 113 | ||
XM_015115891.2 | XP_014971377.1 | 139 | 113 | |||
GenBank mRNA | JV043461.1 | - | 131 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |