Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044738.1 | NP_001038203.1 | 408 | 106 | ||
XM_015143100.2 | XP_014998586.1 | 834 | 106 | |||
XM_015143101.2 | XP_014998587.1 | 602 | 106 | |||
XM_015143102.2 | XP_014998588.1 | 612 | 106 | |||
GenBank mRNA | JU335792.1 | - | 587 | 106 | ||
JU476671.1 | - | 489 | 106 | |||
JV047845.1 | - | 454 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |