Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STPG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190958.2 | NP_001177887.2 | 144 | 110 | ||
XM_015112270.2 | XP_014967756.2 | 674 | 110 | |||
XM_015112271.2 | XP_014967757.2 | 274 | 110 | |||
XM_028831639.1 | XP_028687472.1 | 274 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |