Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pttg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022391.2 | NP_071786.1 | 1 - 2 | 368 | 81 | |
XM_006246142.1 | XP_006246204.1 | 2 - 3 | 179 | 81 | ||
XM_006246143.2 | XP_006246205.1 | 2 - 3 | 376 | 81 | ||
XM_006246144.1 | XP_006246206.1 | 2 - 3 | 473 | 81 | ||
XM_006246145.2 | XP_006246207.1 | 2 - 3 | 179 | 81 | ||
XM_008767643.1 | XP_008765865.1 | 2 - 3 | 128 | 81 | ||
GenBank mRNA | BC158551.1 | - | 2 - 3 | 478 | 9032805 | 81 |
U73030.1 | - | 1 - 2 | 380 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |