Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptger4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032076.3 | NP_114465.3 | 1 - 2 | 873 | 70 | |
XM_006231996.2 | XP_006232058.1 | 2 - 3 | 3442 | 70 | ||
GenBank mRNA | D28860.1 | - | 2 - 3 | 1053 | 70 | |
U94709.1 | - | 1 - 2 | 873 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |