Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053964.1 | NP_446416.1 | 1 - 2 | 1021 | 73 | |
GenBank mRNA | X98170.1 | - | 1 - 2 | 1013 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |