Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035237.1 | NP_001030314.1 | 3 - 4 | 293 | 72 | |
NM_133524.2 | NP_598208.2 | 3 - 4 | 293 | 72 | ||
GenBank mRNA | AJ973227.1 | - | 3 - 4 | 293 | 72 | |
BC078750.1 | - | 3 - 4 | 221 | 7123064 | 72 | |
X54549.1 | - | 3 - 4 | 293 | 72 | ||
X62323.1 | - | 3 - 4 | 194 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |