Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ripk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139342.1 | NP_647558.1 | 2 - 3 | 368 | 70 | |
XM_008770662.1 | XP_008768884.1 | 1 - 2 | 552 | 70 | ||
GenBank mRNA | AF036537.2 | - | 2 - 3 | 368 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |