Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153736.1 | NP_714958.1 | 1 - 2 | 247 | 106 | |
GenBank mRNA | AF526270.1 | - | 1 - 2 | 247 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Inventoried Probe spans exons |